Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfyve21
Entrez Gene ID
68520
Gene Name
zinc finger, FYVE domain containing 21
Gene Aliases
1110013H04Rik, AW558780, C85416
Chromosome Location
Chr.12: 111814170 - 111828388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026752.4 NP_081028.3 2 - 3 256 95
GenBank mRNA AK003661.1 - 2 - 3 153 95
AK139356.1 - - 191 95
AK155300.1 - 2 - 3 257 95
AK171099.1 - 2 - 3 197 95
BC019521.1 - 2 - 3 186 95
BC132248.1 - 2 - 3 156 95
BC132250.1 - 2 - 3 156 95

View Full Product Details