The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163687.1 | NP_001157159.1 | 10 - 11 | 1131 | 93 |
NM_025972.4 | NP_080248.2 | 10 - 11 | 1137 | 93 | |
GenBank mRNA | AK008776.1 | - | 10 - 11 | 1139 | 93 |
AK017141.1 | - | 10 - 11 | 1114 | 93 | |
AK089243.1 | - | 2 - 3 | 93 | ||
AK133485.1 | - | 10 - 11 | 1122 | 93 | |
AK133658.1 | - | 10 - 11 | 1138 | 93 | |
AK146191.1 | - | 10 - 11 | 1125 | 93 | |
BC004572.1 | - | 10 - 11 | 1107 | 93 |