The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007711.3 | NP_031737.1 | 10 - 11 | 2520 | 78 |
NM_173873.1 | NP_776298.1 | 11 - 12 | 2696 | 78 | |
NM_173874.1 | NP_776299.1 | 11 - 12 | 2696 | 78 | |
NM_173876.3 | NP_776301.1 | 10 - 11 | 2241 | 78 | |
GenBank mRNA | AF029347.1 | - | 10 - 11 | 2144 | 78 |
AK141136.1 | - | 11 - 12 | 2634 | 78 | |
AK147364.1 | - | 9 - 10 | 6719 | 78 | |
AK147548.1 | - | 9 - 10 | 6721 | 78 | |
AK147621.1 | - | 9 - 10 | 6721 | 78 | |
AK150688.1 | - | 11 - 12 | 2625 | 78 | |
AK159698.1 | - | 11 - 12 | 2670 | 78 | |
AK161327.1 | - | 11 - 12 | 2624 | 78 | |
AK162639.1 | - | 11 - 12 | 2637 | 78 | |
AK169297.1 | - | 11 - 12 | 2617 | 78 | |
BC057133.1 | - | 10 - 11 | 2174 | 78 | |
X78874.1 | - | 10 - 11 | 2345 | 78 |