Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctsj
Entrez Gene ID
26898
Gene Name
cathepsin J
Gene Aliases
CATP, CatRLP, Ctsp
Chromosome Location
Chr.13: 61000277 - 61005958 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012007.1 NP_036137.1 7 - 8 980 79
GenBank mRNA AF136272.1 - 7 - 8 980 79
AF158182.1 - 6 - 7 920 79
AK005526.1 - 7 - 8 987 79
AK131661.1 - 7 - 8 987 79
BC103769.1 - 7 - 8 976 79

View Full Product Details