The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164789.1 | NP_001158261.1 | 1 - 2 | 1356 | 78 |
NM_010136.3 | NP_034266.2 | 1 - 2 | 1356 | 78 | |
GenBank mRNA | AB031037.1 | - | 1 - 2 | 1328 | 78 |
AF013281.1 | - | 1 - 2 | 54 | 78 | |
AK043327.1 | - | 1 - 2 | 687 | 78 | |
AK089817.1 | - | 1 - 2 | 916 | 78 | |
AK134951.1 | - | - | 710 | 78 | |
AK143454.1 | - | 1 - 2 | 677 | 78 | |
BC094319.1 | - | 1 - 2 | 1201 | 78 |