Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eomes
Entrez Gene ID
13813
Gene Name
eomesodermin homolog (Xenopus laevis)
Gene Aliases
C77258, TBR-2, Tbr2
Chromosome Location
Chr.9: 118478180 - 118486132 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164789.1 NP_001158261.1 5 - 6 1856 83
GenBank mRNA AB031037.1 - 5 - 6 1828 83
AY836552.1 - 2 - 3 82 83

View Full Product Details