The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009455.3 | NP_033481.1 | 2 - 3 | 341 | 82 |
GenBank mRNA | AK090177.1 | - | 2 - 3 | 342 | 82 |
AK148266.1 | - | 2 - 3 | 272 | 82 | |
BC003781.1 | - | 2 - 3 | 258 | 82 | |
X92665.1 | - | 2 - 3 | 303 | 82 |