Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zscan4a
Entrez Gene ID
434555
Gene Name
zinc finger and SCAN domain containing 4A
Gene Aliases
BQ559217
Chromosome Location
Chr.7: 10794241 - 10799065 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033707.1 3 - 4 711 140

View Full Product Details