Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r15
Entrez Gene ID
211223
Gene Name
vomeronasal 2, receptor 15
Gene Aliases
EG211223
Chromosome Location
Chr.5: 109286269 - 109297556 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104626.1 NP_001098096.1 4 - 5 1541 176

View Full Product Details