The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177101.5 | NP_796075.2 | 1 - 2 | 191 | 91 |
GenBank mRNA | AK029471.1 | - | 1 - 2 | 151 | 91 |
AK077292.1 | - | 1 - 2 | 98 | 91 | |
BB614316.1 | - | 1 - 2 | 151 | 91 | |
BC048711.1 | - | 1 - 2 | 83 | 91 | |
BC065807.1 | - | 1 - 2 | 107 | 91 |