The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027942.1 | NP_082218.1 | 1 - 1 | 1462 | 63 |
GenBank mRNA | AK005882.1 | - | 1 - 1 | 1464 | 63 |