The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019750.3 | NP_062724.1 | 2 - 2 | 1824 | 148 |
GenBank mRNA | AF172275.1 | - | 2 - 2 | 1204 | 148 |
AK137391.1 | - | 2 - 2 | 1190 | 148 | |
BC026545.1 | - | 2 - 2 | 1849 | 148 |