Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Jpx
- Entrez Gene ID
-
70252
- Gene Name
- Jpx transcript, Xist activator (non-protein coding)
- Gene Aliases
-
2010000I03Rik, 2510040I06Rik, Enox
- Chromosome Location
-
Chr.X: 103493576 - 103506425 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK137473.1 |
- |
3 - 3 |
473 |
95 |
AK148110.1 |
- |
3 - 3 |
492 |
95 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AV107747.2 |
- |
3 |
588 |
|
Chr. X - 103493770 - 103503237 [+] on Build GRCm38 |
95 |
GenBankmRNA |
AJ421479.1 |
- |
|
|
|
Chr. - - [-] on Build |
95 |
GenBankmRNA |
BY722650.1 |
- |
3 |
472 |
|
Chr. X - 103493576 - 103503332 [+] on Build GRCm38 |
95 |
GenBankmRNA |
BY741299.1 |
- |
3 |
477 |
|
Chr. X - 103493570 - 103503351 [+] on Build GRCm38 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
X (inactive)-specific transcript, opposite strand |
Tsix |
|
-17.9kb
|
Sense |
Chr. - 103431517 - 103484957 on Build GRCm38 |
Downstream |
polycomb group ring finger 6 pseudogene |
Gm9785 |
ENSMUSG00000043661 |
22.0kb
|
Anti-sense |
Chr. - 103525319 - 103525847 on Build GRCm38 |
View Full Product Details