Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam60a
Entrez Gene ID
56306
Gene Name
family with sequence similarity 60, member A
Gene Aliases
Ppcs1, Pptcs1, Tera
Chromosome Location
Chr.6: 148921045 - 148946432 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019643.3 NP_062617.2 4 - 5 516 147
GenBank mRNA AK046519.1 - 4 - 5 423 147
AK145384.1 - 4 - 5 527 147
AK166569.1 - 4 - 5 522 147
BC076599.1 - 4 - 5 516 147
U64033.1 - 4 - 5 605 147

View Full Product Details