The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019643.3 | NP_062617.2 | 2 - 3 | 290 | 112 |
GenBank mRNA | AK046519.1 | - | 2 - 3 | 197 | 112 |
AK145384.1 | - | 2 - 3 | 301 | 112 | |
AK166569.1 | - | 2 - 3 | 296 | 112 | |
BC076599.1 | - | 2 - 3 | 290 | 112 | |
U64033.1 | - | 2 - 3 | 379 | 112 |