Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slitrk1
Entrez Gene ID
76965
Gene Name
SLIT and NTRK-like family, member 1
Gene Aliases
3200001I04Rik
Chromosome Location
Chr.14: 108909988 - 108914239 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199065.2 NP_951020.1 1 - 1 2990 102
GenBank mRNA AK014285.1 - 1 - 1 926 102
AK173296.1 - - 2909 102
BC150756.1 - 1 - 1 2098 102
BC157902.1 - 1 - 1 2098 102

View Full Product Details