Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trcg1
Entrez Gene ID
541610
Gene Name
taste receptor cell gene 1
Gene Aliases
-
Chromosome Location
Chr.9: 57236460 - 57252175 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014398.2 NP_001014420.2 12 - 13 2568 80
GenBank mRNA AY701870.1 - 12 - 13 2568 80
AY701871.1 - 10 - 11 1006 80

View Full Product Details