Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trcg1
Entrez Gene ID
541610
Gene Name
taste receptor cell gene 1
Gene Aliases
-
Chromosome Location
Chr.9: 57236460 - 57252175 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014398.2 NP_001014420.2 4 - 5 1668 97
GenBank mRNA AY701870.1 - 4 - 5 1668 97
AY701871.1 - 3 - 4 252 97

View Full Product Details