Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr37
Entrez Gene ID
207615
Gene Name
WD repeat domain 37
Gene Aliases
-
Chromosome Location
Chr.13: 8802966 - 8871904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039388.2 NP_001034477.1 3 - 4 531 118
NM_001039389.2 NP_001034478.1 3 - 4 452 118
NM_172445.3 NP_766033.1 3 - 4 452 118
GenBank mRNA AK014128.1 - 3 - 4 428 118
AK032582.1 - 3 - 4 414 118
AK036203.1 - 3 - 4 448 118
AK044740.1 - 3 - 4 432 118
AK129254.1 - 3 - 4 408 118
AK146209.1 - 3 - 4 427 118
BB611667.1 - 3 - 4 531 118
BC046236.1 - 2 - 3 279 118
BC048819.1 - 3 - 4 376 118
CJ134699.1 - 3 - 4 453 118

View Full Product Details