Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rps26
- Entrez Gene ID
-
27370
- Gene Name
- ribosomal protein S26
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 128624529 - 128626506 on Build GRCm38
- Gene Symbol
- Gm6654
- Entrez Gene ID
-
626175
- Gene Name
- predicted pseudogene 6654
- Gene Aliases
-
EG626175
- Chromosome Location
-
Chr.6: 146647177 - 146647802 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_038089.1 |
|
1 - 1 |
395 |
96 |
- Gene Symbol
- Med21
- Entrez Gene ID
-
108098
- Gene Name
- mediator complex subunit 21
- Gene Aliases
-
0610007L03Rik, AI449604, D19234, D6Ertd782e, Srb7, Surb7
- Chromosome Location
-
Chr.6: 146642579 - 146650600 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC096641.1 |
- |
1 - 1 |
|
96 |
View Full Product Details