Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Gdf11
Entrez Gene ID
14561
Gene Name
growth differentiation factor 11
Gene Aliases
BMP-11, Bmp11
Chromosome Location
Chr.10: 128884546 - 128891718 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010272.1 NP_034402.1 1 - 1 463 144
GenBank mRNA AF092734.1 - 1 - 1 208 144

View Full Product Details