Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Suv39h1
Entrez Gene ID
20937
Gene Name
suppressor of variegation 3-9 homolog 1 (Drosophila)
Gene Aliases
AI852103, AL022883, DXHXS7466e, H3-K9-HMTase 1, KMT1A, mIS6
Chromosome Location
Chr.X: 8061171 - 8074760 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290716.1 NP_001277645.1 6 - 6 2897 122
NM_011514.2 NP_035644.1 6 - 6 2916 122
GenBank mRNA AF019969.1 - 6 - 6 2744 122

View Full Product Details