The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013692.3 | NP_038720.1 | 4 - 4 | 2981 | 123 |
GenBank mRNA | AF049879.1 | - | 4 - 4 | 2977 | 123 |
BC003316.1 | - | 4 - 4 | 2921 | 123 | |
BY548635.1 | - | 1 - 1 | 381 | 123 | |
Z36270.1 | - | 4 - 4 | 2995 | 123 |