Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sh3d19
- Entrez Gene ID
-
27059
- Gene Name
- SH3 domain protein D19
- Gene Aliases
-
AW011754, Kryn
- Chromosome Location
-
Chr.3: 85970956 - 86130522 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK035963.1 |
- |
1 - 1 |
1250 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
protease, serine 48 |
Prss48 |
Esspl, Gm1019 |
-119.3kb
|
Anti-sense |
Chr. - 85973843 - 86006002 on Build GRCm38 |
Downstream |
ribosomal protein S3A1 |
Rps3a1 |
Rps3a |
8.8kb
|
Anti-sense |
Chr. - 86137940 - 86142668 on Build GRCm38 |
View Full Product Details