Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc8c
Entrez Gene ID
100604
Gene Name
leucine rich repeat containing 8 family, member C
Gene Aliases
AD158, AI326115, E430036I04Rik, fad158
Chromosome Location
Chr.5: 105519386 - 105613018 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK168156.1 - 1 - 1 453 118


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA W85380.1 - 182 Chr. - - [-] on Build 118
GenBankmRNA BY738364.1 - 1 453 Chr. 5 - 105611797 - 105612446 [+] on Build GRCm38 118
GenBankmRNA W81775.1 - 187 Chr. - - [-] on Build 118
GenBankmRNA BM946607.1 - 1 230 Chr. 5 - 105612021 - 105612764 [+] on Build GRCm38 118


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream leucine rich repeat containing 8 family, member B Lrrc8b R75581, Ta-lrrp, mKIAA0231 -121.7kb Sense Chr. - 105415775 - 105490071 on Build GRCm38
Downstream predicted gene, 42166 Gm42166 84.0kb Anti-sense Chr. - 105697029 - 105704089 on Build GRCm38

View Full Product Details