Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- 2810017D21Rik
- Entrez Gene ID
-
72638
- Gene Name
- RIKEN cDNA 2810017D21 gene
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK012752.1 |
- |
1 - 1 |
551 |
75 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BF719779.1 |
- |
1 |
|
|
Chr. 4 - 130709923 - 130710427 [-] on Build GRCm38 |
75 |
GenBankmRNA |
DV075383.1 |
- |
|
388 |
|
Chr. - - [-] on Build |
75 |
GenBankmRNA |
CB850000.2 |
- |
1 |
|
|
Chr. 4 - 130709677 - 130710508 [-] on Build GRCm38 |
75 |
GenBankmRNA |
CB194530.1 |
- |
1 |
481 |
|
Chr. 4 - 130709741 - 130710509 [+] on Build GRCm38 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
pumilio RNA-binding family member 1 |
Pum1 |
AA517475, Pumm, mKIAA0099 |
|
Sense |
Chr. - 130663297 - 130781564 on Build GRCm38 |
Downstream |
small nucleolar RNA, C/D box 85 |
Snord85 |
MBII-251, Z50 |
39.1kb
|
Sense |
Chr. - 130749638 - 130749702 on Build GRCm38 |
View Full Product Details