Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
9130604C24Rik
Entrez Gene ID
77707
Gene Name
RIKEN cDNA 9130604C24 gene
Gene Aliases
AW554730
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK020298.1 - 1 - 1 645 77
AW554730.1 - 1 - 1 77
BG074933.2 - 1 - 1 77


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream processing of precursor 7, ribonuclease P family, (S. cerevisiae) Pop7 0610037N12Rik, AI852017, Rpp20 -242.0 Anti-sense Chr. - 137501439 - 137502429 on Build GRCm38
Downstream GRB10 interacting GYF protein 1 Gigyf1 Perq1, mKIAA4110 6.6kb Sense Chr. - 137510258 - 137527935 on Build GRCm38

View Full Product Details