The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005860.2 | NP_001005860.1 | 3 - 4 | 297 | 99 |
GenBank mRNA | AY397673.1 | - | 3 - 4 | 297 | 99 |
BC104370.1 | - | 3 - 4 | 297 | 99 | |
BC104371.2 | - | 3 - 4 | 296 | 99 |