Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Magea1
Entrez Gene ID
17137
Gene Name
melanoma antigen, family A, 1
Gene Aliases
Mage-a1
Chromosome Location
Chr.X: 155088519 - 155089790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020015.2 NP_064399.1 1 - 1 544 108
GenBank mRNA BC116904.1 - 1 - 1 544 108

Gene Symbol
Magea2
Entrez Gene ID
17138
Gene Name
melanoma antigen, family A, 2
Gene Aliases
Mage-a2
Chromosome Location
Chr.X: 155027201 - 155033313 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020016.1 NP_064400.1 4 - 4 781 108
GenBank mRNA BC106842.2 - - 681 108

View Full Product Details