Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mnda
Entrez Gene ID
381308
Gene Name
myeloid cell nuclear differentiation antigen
Gene Aliases
A730048F03, Ifi205b, ifi-205-B
Chromosome Location
Chr.1: 173896341 - 173913046 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033450.4 NP_001028622.1 5 - 6 1291 104
NM_001301745.1 NP_001288674.1 5 - 6 1285 104
GenBank mRNA AK152278.1 - 5 - 6 1159 104
AK152683.1 - 5 - 6 1159 104
AK152786.1 - 5 - 6 1168 104
BC132314.1 - 5 - 6 1112 104
BC132316.1 - 5 - 6 1112 104
BC145397.1 - 5 - 6 1106 104
M74123.1 - 5 - 6 1146 104
S62227.1 - 5 - 6 1146 104

Gene Symbol
Ifi205
Entrez Gene ID
226695
Gene Name
interferon activated gene 205
Gene Aliases
D3, D3cDNA, Ifi205a
Chromosome Location
Chr.1: 174011998 - 174031755 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172648.3 NP_766236.2 5 - 6 1127 104
GenBank mRNA AK040191.1 - 5 - 6 1057 104
BC040425.1 - 5 - 6 1127 104
CT010211.1 - 4 - 5 975 104
M74124.1 - 4 - 5 908 104

View Full Product Details