Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r41
Entrez Gene ID
100042848
Gene Name
vomeronasal 2, receptor 41
Gene Aliases
-
Chromosome Location
Chr.7: 8137905 - 8161551 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105073.1 NP_001098543.1 4 - 5 1529 105

Gene Symbol
Vmn2r40
Entrez Gene ID
100042781
Gene Name
vomeronasal 2, receptor 40
Gene Aliases
-
Chromosome Location
Chr.7: 8907734 - 8931402 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105072.1 NP_001098542.1 4 - 5 1529 105

Gene Symbol
Vmn2r48
Entrez Gene ID
625580
Gene Name
vomeronasal 2, receptor 48
Gene Aliases
EG625580
Chromosome Location
Chr.7: 9927624 - 9953585 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105152.1 NP_001098622.1 4 - 5 1538 105

View Full Product Details