Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rhox3g
- Entrez Gene ID
-
546294
- Gene Name
- reproductive homeobox 3G
- Gene Aliases
-
OTTMUSG00000017157, Rhox3.7, Rhox3f, Rhox3g-ps
- Chromosome Location
-
Chr.X: 37623431 - 37628139 on Build GRCm38
- Gene Symbol
- Rhox3h
- Entrez Gene ID
-
434758
- Gene Name
- reproductive homeobox 3H
- Gene Aliases
-
EG434758, Rhox3.8, Rhox3g
- Chromosome Location
-
Chr.X: 37657688 - 37668764 on Build GRCm38
View Full Product Details