Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpina3a
Entrez Gene ID
74069
Gene Name
serine (or cysteine) peptidase inhibitor, clade A, member 3A
Gene Aliases
-
Chromosome Location
Chr.12: 104106704 - 104121897 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167705.1 NP_001161177.1 1 - 2 202 86

View Full Product Details