Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hoxd8
Entrez Gene ID
15437
Gene Name
homeobox D8
Gene Aliases
4921540P06Rik, AI047735, Hox-4.3, Hox-5.4
Chromosome Location
Chr.2: 74704615 - 74707933 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290731.1 NP_001277660.1 2 - 3 260 86
NM_008276.3 NP_032302.2 1 - 2 632 86
GenBank mRNA AK016033.1 - 1 - 2 361 86
BX513411.1 - 2 - 3 284 86

View Full Product Details