The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198661.3 | NP_941063.1 | 2 - 3 | 138 | 88 |
GenBank mRNA | AY573562.1 | - | 2 - 3 | 138 | 88 |
BC052839.1 | - | 2 - 3 | 138 | 88 |