Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ssxb10
- Entrez Gene ID
-
385312
- Gene Name
- synovial sarcoma, X member B, breakpoint 10
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 8327424 - 8336235 on Build GRCm38
- Gene Symbol
- Ssxb2
- Entrez Gene ID
-
387132
- Gene Name
- synovial sarcoma, X member B, breakpoint 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 8454343 - 8461726 on Build GRCm38
- Gene Symbol
- Ssxb8
- Entrez Gene ID
-
631002
- Gene Name
- synovial sarcoma, X member B, breakpoint 8
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 8684843 - 8690883 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY244799.1 |
- |
- |
|
115 |
View Full Product Details