Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekhf2
Entrez Gene ID
71801
Gene Name
pleckstrin homology domain containing, family F (with FYVE domain) member 2
Gene Aliases
1110070J07Rik, AA673237, ZFYVE18
Chromosome Location
Chr.4: 10988662 - 11007619 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175175.4 NP_780384.1 2 - 2 1049 93
GenBank mRNA AK033666.1 - 2 - 2 1043 93
AK154634.1 - 2 - 2 1040 93
BC039276.1 - 2 - 2 1000 93

View Full Product Details