The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253738.1 | NP_001240667.1 | 7 - 8 | 870 | 62 |
NM_001253740.1 | NP_001240669.1 | 7 - 8 | 870 | 62 | |
NM_022314.3 | NP_071709.3 | 8 - 9 | 853 | 62 | |
GenBank mRNA | AK088111.1 | - | 7 - 8 | 730 | 62 |
AK088902.1 | - | 7 - 8 | 728 | 62 | |
AK145238.1 | - | 7 - 8 | 727 | 62 | |
AK145243.1 | - | 7 - 8 | 727 | 62 | |
AK145804.1 | - | 7 - 8 | 727 | 62 | |
AK146349.1 | - | 7 - 8 | 700 | 62 | |
AK146557.1 | - | 7 - 8 | 852 | 62 | |
AK151441.1 | - | 7 - 8 | 752 | 62 | |
AK151500.1 | - | 7 - 8 | 727 | 62 | |
AK151571.1 | - | 7 - 8 | 726 | 62 | |
AK151578.1 | - | 7 - 8 | 727 | 62 | |
AK151595.1 | - | 7 - 8 | 699 | 62 | |
AK151602.1 | - | 7 - 8 | 727 | 62 | |
AK151991.1 | - | 7 - 8 | 726 | 62 | |
AK152260.1 | - | 7 - 8 | 726 | 62 | |
AK152659.1 | - | 7 - 8 | 699 | 62 | |
AK153031.1 | - | 7 - 8 | 699 | 62 | |
AK160011.1 | - | 7 - 8 | 752 | 62 | |
AK165428.1 | - | 7 - 8 | 726 | 62 | |
AK166836.1 | - | 7 - 8 | 714 | 62 | |
AK167268.1 | - | 7 - 8 | 704 | 62 | |
AK167534.1 | - | 7 - 8 | 732 | 62 | |
AK167581.1 | - | 7 - 8 | 727 | 62 | |
AK168843.1 | - | 7 - 8 | 695 | 62 | |
AK169102.1 | - | 7 - 8 | 698 | 62 | |
AK169245.1 | - | 7 - 8 | 703 | 62 | |
AK169821.1 | - | 7 - 8 | 744 | 62 | |
BC029186.1 | - | 8 - 9 | 810 | 62 | |
BC055407.1 | - | - | 3996 | 62 | |
BC092045.1 | - | 7 - 8 | 703 | 62 | |
CX237327.1 | - | 7 - 8 | 692 | 62 | |
X53753.1 | - | 7 - 8 | 738 | 62 |