Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB17
Entrez Gene ID
101121217
Gene Name
ankyrin repeat and SOCS box containing 17
Gene Aliases
-
Chromosome Location
Chr.1: 56108949 - 56129919 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002089.4 XP_004002138.1 2 - 3 677 95

View Full Product Details