Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HARBI1
Entrez Gene ID
101114808
Gene Name
harbinger transposase derived 1
Gene Aliases
-
Chromosome Location
Chr.15: 82846757 - 82856232 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016440.3 XP_004016489.1 2 - 3 902 61
XM_012096345.3 XP_011951735.1 2 - 3 900 61

View Full Product Details