Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPAT4
Entrez Gene ID
101117217
Gene Name
glycerol-3-phosphate acyltransferase 4
Gene Aliases
AGPAT6
Chromosome Location
Chr.26: 39880251 - 39910648 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021798.4 XP_004021847.1 2 - 3 1057 87

View Full Product Details