Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRSS42
Entrez Gene ID
101115659
Gene Name
serine protease 42
Gene Aliases
PRSS42P
Chromosome Location
Chr.19: 54626018 - 54630685 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957977.1 XP_027813778.1 1 - 2 200 62

View Full Product Details