Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRM1
Entrez Gene ID
101111204
Gene Name
mitochondrial rRNA methyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.11: 50990663 - 50998790 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012434.4 XP_004012483.2 1 - 2 570 72

View Full Product Details