Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHODL
Entrez Gene ID
101115923
Gene Name
chondrolectin
Gene Aliases
-
Chromosome Location
Chr.1: 149754580 - 149778198 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002816.4 XP_004002865.1 2 - 3 442 62

View Full Product Details