Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHM3
Entrez Gene ID
101119021
Gene Name
pleckstrin homology domain containing M3
Gene Aliases
-
Chromosome Location
Chr.2: 224019254 - 224233401 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004870.4 XP_004004919.2 3 - 4 1998 61

View Full Product Details