Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSPG4
Entrez Gene ID
101105108
Gene Name
chondroitin sulfate proteoglycan 4
Gene Aliases
-
Chromosome Location
Chr.18: 31031152 - 31066707 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957164.1 XP_027812965.1 4 - 5 4387 59

View Full Product Details