Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCX
Entrez Gene ID
101109861
Gene Name
doublecortin
Gene Aliases
-
Chromosome Location
Chr.X: 132029007 - 132143581 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963482.1 XP_027819283.1 3 - 4 824 74

View Full Product Details