Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLR10
Entrez Gene ID
554255
Gene Name
toll like receptor 10
Gene Aliases
-
Chromosome Location
Chr.6: 63575101 - 63577579 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135925.1 NP_001129397.1 1 - 1 2322 71
GenBank mRNA AM981308.1 - 1 - 1 2322 71

View Full Product Details