Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICE1
Entrez Gene ID
101107734
Gene Name
interactor of little elongation complex ELL subunit 1
Gene Aliases
-
Chromosome Location
Chr.16: 74033908 - 74103190 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027980180.1 XP_027835981.1 12 - 13 1275 63

View Full Product Details