Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRN1L
Entrez Gene ID
101116419
Gene Name
neuritin 1 like
Gene Aliases
-
Chromosome Location
Chr.14: 38156998 - 38157727 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977910.1 XP_027833711.1 2 - 3 195 58

View Full Product Details